missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Osteoprotegerin/TNFRSF11B Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 3 publications
POA
Specifications
Antigen | Osteoprotegerin/TNFRSF11B |
---|---|
Clone | 110511 |
Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
Osteoprotegerin/TNFRSF11B Monoclonal specifically detects Osteoprotegerin/TNFRSF11B in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
Osteoprotegerin/TNFRSF11B | |
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
OCIF, OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TNFRSF11B, TR1, tumor necrosis factor receptor superfamily, member 11b | |
TNFRSF11B | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
110511 | |
Western Blot, ELISA | |
Unconjugated | |
Rat | |
Mouse | |
Q6PI12 | |
4982 | |
Mouse myeloma cell line NS0-derived recombinant mouse Osteoprotegerin/TNFRSF11B Gln21-Leu401 (Gln138Arg) Accession # Q6PI12 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse Osteoprotegerin/TNFRSF11B in ELISAs and Western blots. In Western blots, this antibody shows 100% cross-reactivity with recombinant human (rh) Osteoprotegerin and no cross-reactivity with rh4-1BB, recombinant mouse (rm) CD27, rmCD40, rhDR3, rhDR6, rmFas, rmGITR, rhHVEM, rhLT beta R, rmRANK, rmTNF RI, or rmTNF RII. |
For Research Use Only