missing translation for 'onlineSavingsMsg'
Learn More

Osteopontin, eFluor 660, clone: 2F10, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-9096-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15510077

  • £81.61 / 25 tests


Estimated Shipment date
from Supplier 21-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 2F10 monoclonal antibody recognizes human osteopontin (OPN), a secreted glycophosphoprotein expressed by multiple cell types, including leukocytes, fibroblasts, dendritic cells, macrophages, myoblasts, bone-forming cells, and epithelial cells. OPN is an extracellular matrix protein with multiple roles, including bone resorption and calcification, immune system regulation, wound healing, and tumorigenesis. Thrombin cleavage of OPN allows binding of integrin receptors, expressed by immune cells, and subsequent modulation of cell adhesion, migration, and survival. OPN is expressed in bones, teeth, placenta, blood vessels, kidney, and various tumors. Osteopontin has been identified as a potential therapeutic target in the treatment of autoimmune disease, cancer metastasis, bone mineralization diseases, and osteoporosis.Applications Reported: This 2F10 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 2F10 antibody has been pre-titrated and tested. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Osteopontin
2F10
eFluor 660
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
bone sialoprotein I (BSP-1)
IgG1
RUO
Primary
Human
Videos