missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ OSMR Recombinant Mouse Protein, His Tag

Expressed from DNA sequence encoding Mouse OSMR extracellular domain (Met 1-Leu 738) fused to polyhistidine tag at C-terminus

Brand:  Sino Biological™ 50500M08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15308866

  • £57.00 / 5µg


Estimated Shipment
13-06-2018
Add to basket

Description

Description

  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
18414
≥90% by SDS-PAGE
Mouse
<1 EU/μg
OSMR
Cytokines and Receptors, CD Proteins
OSMR Recombinant Mouse Protein
Lyophilized
83.7kd
5μg
Truncated, Recombinant
Oncostatin M Receptor
Assay Standard
Mammalian
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.