missing translation for 'onlineSavingsMsg'
Learn More

Orotic acid monohydrate, 98%, Thermo Scientific Chemicals

Catalog Number p-3758611
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
50887-69-9
C5H4N2O4
156.10
MFCD00006027
PXQPEWDEAKTCGB-UHFFFAOYSA-N
orotic acid monohydrate, orotic acid hydrate, 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate, lactinium, oroticum acidum, 2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid, orotc acd hydrate?, uracil-6-carboxylic acid monohydrate, uracil-4-carboxylic acid hydrate
16218504
2,4-dioxo-1H-pyrimidine-6-carboxylic acid;hydrate
OC(=O)C1=CC(=O)NC(=O)N1
This item is not returnable. View return policy
50887-69-9
C5H4N2O4
156.10
MFCD00006027
PXQPEWDEAKTCGB-UHFFFAOYSA-N
orotic acid monohydrate, orotic acid hydrate, 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate, lactinium, oroticum acidum, 2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid, orotc acd hydrate?, uracil-6-carboxylic acid monohydrate, uracil-4-carboxylic acid hydrate
16218504
2,4-dioxo-1H-pyrimidine-6-carboxylic acid;hydrate
OC(=O)C1=CC(=O)NC(=O)N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 334°C
Color White
Loss on Drying 12.0% max. (1 g, 130°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 5 g
Beilstein 25, 253
Merck Index 15, 6972
Solubility Information Solubility in water: 1.3g/L (20 c). Other solubilities: insoluble in chloroform
Formula Weight 174.11
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Orotic acid monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • Orotic acid monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'