Learn More
Orlistat, Tocris Bioscience™
Pancreatic, gastric and carboxylester lipase inhibitor; antiobesity and antihypercholesterolemic activity
Brand:  Tocris (Bio-Techne) 354010
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13443164
  • £67.50 / 10mg
Estimated Shipment
27-06-2019
Description
Description
Hypolipemic pancreatic, gastric and carboxylester lipase inhibitor. Exhibits no activity at phospholipase A2, liver esterase, trypsin and chymotrypsin. Inhibits the thioesterase domain of fatty acid synthase, leading to cell cycle arrest at the G1/S boundary in vitro. Prevents the absorption of approximately one third of fat from food and exhibits progastrokinetic, antiobesity and antihypercholesterolemic activity in vivo.
Specifications
Chemical Identifiers
96829-58-2
495.745
orlistat, tetrahydrolipstatin, xenical, alli, orlipastat, --tetrahydrolipstatin, orlipastatum, orlipastatum inn-latin, thlp, tetrahydrolipastatin
[(2S)-1-[(2S,3S)-3-hexyl-4-oxooxetan-2-yl]tridecan-2-yl] (2S)-2-formamido-4-methylpentanoate
C29H53NO5
AHLBNYSZXLDEJQ-FWEHEUNISA-N
3034010
CCCCCCCCCCCC(CC1C(C(=O)O1)CCCCCC)OC(=O)C(CC(C)C)NC=O
Specifications
Orlistat
96829-58-2
orlistat, tetrahydrolipstatin, xenical, alli, orlipastat, --tetrahydrolipstatin, orlipastatum, orlipastatum inn-latin, thlp, tetrahydrolipastatin
CCCCCCCCCCCC(CC1C(C(=O)O1)CCCCCC)OC(=O)C(CC(C)C)NC=O
495.745
495.73
10mg
C29H53NO5
AHLBNYSZXLDEJQ-FWEHEUNISA-N
[(2S)-1-[(2S,3S)-3-hexyl-4-oxooxetan-2-yl]tridecan-2-yl] (2S)-2-formamido-4-methylpentanoate
3034010
>99%
Videos