missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Nitrate Electrodes

Measure nitrate in aqueous solutions simply, accurately and economically with these electrodes.

Brand:  Thermo Scientific™ 970701

244.33 GBP valid until 2024-04-30
Use promo code "21656" to get your promotional price.



Code : 59

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10374571

  • £265.89 / Each
In Stock
to see stock.
Add to basket

Description

Description

Measure nitrate in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Nitrate Electrodes. A separate reference electrode is unnecessary with this electrode, making it convenient to use with small sample volumes.

  • Calibration standards catalog number: 920706 — 0.1M NaNO3; 920707 — 1000ppm N; 930707 — 100ppm N
  • Required ISA catalog number: 930711 or 930710/nitrate ISS
Specifications

Specifications

Replacement Module for Nitrate Sure-Flow Combination Electrode 9707BNWP; 1/pk.
Free-flowing Sure-Flow™ junction assures stable
Combination Ion Selective Electrode
drift-free potentials
ISE Meters
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers