Learn More
Thermo Scientific™ Orion™ Electrode Extension Cables
Extend the length of an electrode cable by 15′ (4.57m) when using the Thermo Scientific™ Orion™ Electrode Extension Cables.
Brand:  Thermo Scientific™ 910028
UNSPSC : 41110000
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11712797
  • £198.45 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications Description & Specifications
Specifications
917005, 917006, and 917007 ATC Probes
15 ft. Extension Cable, 8 Pin DIN connector for 917005, 917006, and 917007 ATC probes
917005, 917006, and 917007 ATC probes
8 Pin DIN
4.57m
Electrode Extension Cable