missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Orion™ Chlorine Electrode, with waterproof BNC connector

Measure total residual chlorine quickly and accurately with these chlorine electrodes.

Brand:  Thermo Scientific™ 9770BNWP

468.11 GBP valid until 2024-04-30
Use promo code "21656" to get your promotional price.



Code : 59

Additional Details : Weight : 1.00000kg

Product Code. 10448912

  • £566.20 / Each
Estimated Shipment: 13-05-2024
to see stock.
Add to basket

Description

Description

Measure total residual chlorine quickly and accurately with the Thermo Scientific™ Orion™ Chlorine Electrodes. All forms of total residual chlorine are measured.

The method permits rapid analysis to be done at the sampling site, eliminating erroneous low readings due to Cl2 loss.

  • Method requires addition of two reagents to the sample before measurement
  • Electrode has a slope of about +29mV per decade increase in concentration
  • Calibration standards catalog number: 100ppm as Cl2 (977007)
  • Required ISA catalog number: 977010 iodide reagent; 977011 acid reagent
Specifications

Specifications

Waterproof BNC
Waterproof BNC connector
ISE Meters
Combination Ion Selective Electrode
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers