Learn More
Thermo Scientific™ Orion™ ROSS™ Sure-Flow™ pH Electrode with Sure-flow junction, BNC connector
Measure general pH samples as well as viscous or colloidal samples with the Thermo Scientific™ Orion™ 8172BNWP ROSS™ Sure-Flow™ pH Electrode.
Brand: Thermo Scientific™ 8172BNWP
Code : 59
Additional Details : Weight : 0.46980kg
Description
Measure general pH samples as well as viscous or colloidal samples with the free-flowing sleeve junction design of the Thermo Scientific™ Orion™ 8172BNWP ROSS™ Sure-Flow™ pH Electrode.
This general purpose pH electrode offers superior performance for dirty, difficult samples such as soils, sludges, colloids, viscous materials and organics. Junction cleans easily between measurements; pressing down on the cap allows filling solution to flow through the junction carries away any contamination that may interfere with measurement.- Material: Glass body
- pH range: 0 to 14
- Temperature range: 0° to 100°C
- Internal reference: ROSS
- Junction: Sure-Flow
- Dimensions: 120 × 12mm
- pH precision: 0.01
- Fill solution: 3M KCl, Orion Cat. No 810007
- Cap diameter: 16mm
Includes:
- Detachable bulb guard, two bottles of Orion ROSS electrode fill solution
Specifications
Orion ROSS Sure-Flow pH electrode | |
Sure-Flow | |
0 to 14 | |
Combination pH Electrode, glass body, BNC waterproof connector | |
Biological/Pharmaceutical –TRIS buffer, proteins, enzymes; Emulsions – Foods, cosmetics, oils; Emulsions – Petroleum products, paint; General Purpose – Most sample types; High Ionic Strength – Acids, bases, brines, pH > 12 or pH < 2; Low Ionic Strength – | |
Detachable bulb guard, 2 x 60mL bottles of ORion ROSS electrode fill solution | |
0.01 | |
16 mm |
WPBNC | |
Glass Body | |
0°C to 100°C | |
3M KCl (810007) | |
Standard pH Meter, BNC connector | |
ROSS | |
32°F to 212°F |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.