missing translation for 'onlineSavingsMsg'
Learn More

Org 24598 lithium salt, Tocris Bioscience™

Selective GlyT1 inhibitor

Brand:  Tocris (Bio-Techne) 403550

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13453994

  • £704.13 / 50mg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective inhibitor of the glial glycine transporter (GlyT1) (pIC50 values are 6.9, <4 and <3 for GlyT1, GABA transporter and Glyt2 respectively).
Specifications

Chemical Identifiers

722456-08-8
373.3
org 24598 lithium salt, chembl61861, n-methyl-n-3r-3-phenyl-3-4-trifluoromethyl phenoxy propyl-glycine lithium salt, r---n-methyl-n-3-4-trifluoromethyl phenoxy-3-phenyl-propyl glycine lithium salt, org 24598 lithium salt hplc , solid, lithium 1+ 2-methyl 3r-3-phenyl-3-4-trifluoromethyl phenoxy propyl amino acetate
lithium;2-[methyl-[(3R)-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]amino]acetate
C19H19F3LiNO3
VMQXVSNARQMSDL-UNTBIKODSA-M
16219807
[Li+].CN(CCC(C1=CC=CC=C1)OC2=CC=C(C=C2)C(F)(F)F)CC(=O)[O-]

Specifications

Org 24598 lithium salt
C19H19F3LiNO3
VMQXVSNARQMSDL-UNTBIKODSA-M
lithium;2-[methyl-[(3R)-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]amino]acetate
16219807
>98%
722456-08-8
org 24598 lithium salt, chembl61861, n-methyl-n-3r-3-phenyl-3-4-trifluoromethyl phenoxy propyl-glycine lithium salt, r---n-methyl-n-3-4-trifluoromethyl phenoxy-3-phenyl-propyl glycine lithium salt, org 24598 lithium salt hplc , solid, lithium 1+ 2-methyl 3r-3-phenyl-3-4-trifluoromethyl phenoxy propyl amino acetate
[Li+].CN(CCC(C1=CC=CC=C1)OC2=CC=C(C=C2)C(F)(F)F)CC(=O)[O-]
373.3
373.3
50mg
Videos
Documents