Learn More
Tocris Bioscience™ Org 12962 hydrochloride
Selective 5-HT2C agonist
Brand:  Tocris Bioscience™ 358550
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13443244
  • £442.02 / 50mg
Estimated Shipment
28-06-2019
Description
Description
Selective 5-HT2C receptor agonist (pEC50 values are 7.01, 6.38 and 6.28 for 5-HT2C, 5-HT2A and 5-HT2B respectively). Displays antiaversive effects in a rat model of panic-like anxiety.
Specifications
Specifications
Org 12962 hydrochloride
>99
50mg
Org 12962 hydrochloride
302.12