missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Orcein, pure
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Absorption Ratio | 0.98 to 1.18 |
Beilstein | 06, 885 |
Merck Index | 15, 6958 |
Infrared Spectrum | Authentic |
Lambda Max | 575 to 582nm |
Formula Weight | 500.51 |
Grade | Pure |
Packaging | Glass bottle |
Color | Black/Purple to Brown |
Quantity | 100 g |
Show More |
Safety and Handling
Product Identifier
- Orcein
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'