missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals orange II

Catalog Number p-3756583
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
633-96-5
C16H11N2NaO4S
350.324
MFCD00011657
IGQFKZCLTLAWLO-UHFFFAOYSA-M
Acid Orange 7, C.I. 15510, p-(2-Hydroxy-1-naphthylazo)benzenesulfonic acid, sodium salt
44135675
sodium;4-[2-(2-oxonaphthalen-1-ylidene)hydrazinyl]benzenesulfonate
C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=C(C=C3)S(=O)(=O)[O-].[Na+]
This item is not returnable. View return policy
633-96-5
C16H11N2NaO4S
350.324
MFCD00011657
IGQFKZCLTLAWLO-UHFFFAOYSA-M
Acid Orange 7, C.I. 15510, p-(2-Hydroxy-1-naphthylazo)benzenesulfonic acid, sodium salt
44135675
sodium;4-[2-(2-oxonaphthalen-1-ylidene)hydrazinyl]benzenesulfonate
C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=C(C=C3)S(=O)(=O)[O-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 164.0°C
Absorption Ratio 0.93 to 0.98 (P-15/P+15)
Bio. Stain Comm. Certification Pass
Dye Content 85% min.
Beilstein 16, 274
Merck Index 15, 6953
Solubility Information Solubility in water: 116g/L (30°C).
Infrared Spectrum Authentic
Lambda Max 480 to 486nm (c=0.01 g/l, H2O)
Formula Weight 350.32
Grade Pure
Packaging Glass bottle
Color Blue to Green
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Orange II, Pure
Show More Show Less
compliance-icons
Product Identifier
  • Orange(II)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'