missing translation for 'onlineSavingsMsg'
Learn More

Orange G, Electrophoresis Grade, Thermo Scientific Chemicals

Catalog Number p-7042498
28.88 GBP valid until 2024-04-26
Use promo code "21687" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
1 kg
This item is not returnable. View return policy
1936-15-8
C16H10N2Na2O7S2
452.36
MFCD00012457
AEOVEGJBKQQFOP-DDVLFWKVSA-L
C.I. 16230; Acid Orange 10
131850556
[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=CC=CC=C3)C(=O)C=C2)C(=C1)S([O-])(=O)=O
This item is not returnable. View return policy
1936-15-8
C16H10N2Na2O7S2
452.36
MFCD00012457
AEOVEGJBKQQFOP-DDVLFWKVSA-L
C.I. 16230; Acid Orange 10
131850556
[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=CC=CC=C3)C(=O)C=C2)C(=C1)S([O-])(=O)=O

Collagen stain for connective tissue

Collagen stain for connective tissue

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Beilstein 4120705
Solubility Information Soluble in water or methyl cellosolve; Slightly soluble in ethanol
IUPAC Name disodium (8Z)-7-oxo-8-(2-phenylhydrazin-1-ylidene)-7,8-dihydronaphthalene-1,3-disulfonate
Formula Weight 452.38
Grade Electrophoresis
Color Orange to Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Orange G

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'