missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR4M2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563624
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NELDSYFCDI TQVVRIACAN TFPEELV Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 93%.
OR4M2 Polyclonal antibody specifically detects OR4M2 in Human samples. It is validated for Immunohistochemistry.Specifications
OR4M2 | |
Polyclonal | |
Unconjugated | |
OR4M2 | |
Olfactory receptor 4M2; olfactory receptor family 4 subfamily M member 2; olfactory receptor OR15-3; olfactory receptor, family 4, subfamily M, member 2; OR15-3; OR4M2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
390538 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NGB6 | |
OR4M2 | |
Recombinant protein corresponding to Human OR4M2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only