missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel™ Optima™ d-6 GC Capillary Column

Brand:  Macherey Nagel™ 726465.25

UNSPSC : 41115710

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10698991

  • £810.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

Ideal for MSD and PND Detectors
0.4mm
25m
Optima® d-6
0.2mm
GC Capillary Column
0.2μm
GC
Optima® d-6
1Each