missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel™ Optima™ d-6 GC Capillary Column

Brand:  Macherey Nagel™ 726482.30

UNSPSC : 41115710

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10690142

  • £835.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

Ideal for MSD and PND Detectors
0.5mm
30m
Optima® d-6
0.32mm
GC Capillary Column
1μm
GC
Optima® d-6
1Each