Learn More
Grant Instruments™ Optima Bridge Plate
View and access thermostat on heating and cooling circulators. Grant Instruments™ Optima Bridge Plate is for use with Grant refrigeration units, heating and cooling circulators.
Brand:  Grant Instruments™ G-BTS
Code : 57
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12757789
  • £118.25 / Each
Estimated Shipment
14-02-2018
Add to basket
Description
Description
  • For use with Grant refrigeration units, heating and cooling circulators
  • Ensures visibility/accessibility of the thermostat whilst optimising bench space
Specifications
Specifications
G Optima Heating Circulator ST5 tank
Videos