Learn More
Grant Instruments™ Optima Bridge Plate
View and access thermostat on heating and cooling circulators. Grant Instruments™ Optima Bridge Plate is for use with Grant refrigeration units, heating and cooling circulators.
Brand:  Grant Instruments™ T-BSL
Code : 57
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12727799
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
View and access thermostat on heating and cooling circulators. Grant Instruments™ Optima Bridge Plate is for use with Grant refrigeration units, heating and cooling circulators.
  • For use with Grant refrigeration units, heating and cooling circulators
  • Ensures visibility/accessibility of the thermostat whilst optimising bench space
Specifications
Specifications
T Optima heating circulator models to S12, 18, 26, 38 tanks
Stainless Steel