Learn More
Grant Instruments™ Optima Bridge Plate
View and access thermostat on heating and cooling circulators. Grant Instruments™ Optima Bridge Plate is for use with Grant refrigeration units, heating and cooling circulators.
Brand:  Grant Instruments™ G-BTL
UNSPSC : 41103715
Code : 57
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12767789
  • £130.00 / Each
Estimated Shipment
02-10-2017
Add to basket
Description
Description
  • For use with Grant refrigeration units, heating and cooling circulators
  • Ensures visibility/accessibility of the thermostat whilst optimising bench space
Specifications
Specifications
G Optima Heating Circulator models to ST12, 18, 26 and 38