missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Optima™ 5 High Performance GC Capillary Column

Brand:  Macherey-Nagel™ 726099.50

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10536511

  • £845.00 / Each
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

0.25 mm
High Performance GC Capillary Column
Optima™ 5
340°C
50 m
G27
0.4 mm
Gas Chromatography (GC)
0.5 μm
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers