missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Optima™ 17 GC Capillary Column

Brand:  Macherey-Nagel™ 726351.30

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10689571

  • £545.00 / Each
Estimated Shipment: 05-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

0.32 mm
GC Capillary Column
Optima™ 17
320°C to 340°C
30 m
G1, G2, G38
0.5 mm
Steroids Analysis, Pesticide Analysis, Drug Analysis
0.25 μm
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers