missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OneComp eBeads™ Compensation Beads

Spherical particles that can be stained with individual fluorochrome-conjugated antibodies for use as single-color compensation controls. OneComp eBeads react with antibodies of mouse, rat and hamster origin, and are immunoglobulin light chain-independent. 1 drop (50μL)/test

Brand:  Invitrogen™ 01-1111-42

173.65 GBP valid until 2024-12-31
Use promo code "22910" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 15242448

  • £200.00 / 100 tests
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!

Alert:

Contains Sodium azide


Description

Description

  • Each drop of beads contains both a positive population that will capture any mouse, rat or hamster antibody and a negative population that will not react with antibody
  • This bimodal distribution can be used for single-color compensation controls in multicolor flow cytometry experiments
  • Cross-reacts to some antibodies of rabbit origin, but have not been extensively tested for this reactivity
  • Designed for use in compensation with all fluorochromes excited by blue (488nm), green (532nm), yellow-green (561nm), and red (633 to 635nm) lasers
  • Compatible with eFluor™ 450 but is not optimized for compensation of other fluorochromes excited by a violet (405nm) laser
Specifications

Specifications

100 Tests
Compensation Beads
eBeads™
Flow Cytometer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.