missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSMR, PE, clone: AN-V2, eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 12-1303-41
Code : NEW
Additional Details : Weight : 0.23750kg
Description
This monoclonal AN-V2 antibody reacts with the human oncostatin M (OSM) receptor. OSM, which belongs to the interleukin-6 cytokine family, is produced by T cells, activated monocytes, and dendritic cells. In humans, OSM can bind two unique receptor complexes. The type I OSM receptor is composed of the LIFR associated with gp130, and recognizes both LIF and OSM. The type II OSM receptor, which consists of gp130 and the OSMR subunit, specifically binds the cytokine, leading to activation of the JAK/STAT signaling pathway. Reports have also indicated a soluble form of the OSM receptor, which this antibody also recognizes. The OSM receptor is expressed in the bone marrow, endothelium, liver, lung, and numerous tumor cell lines.The AN-V2 monoclonal antibody recognizes the type II (beta) OSM receptor.
Specifications
Oncostatin M Receptor | |
AN-V2 | |
PE | |
Conjugated | |
Murine | |
25 Tests | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
Monoclonal |
Flow Cytometry | |
5μL (0.5μg)/test | |
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer | |
OSMR | |
IgG1 | |
RUO | |
Primary | |
Human |
For Research Use Only.