Learn More
Diba Omnifit™ “T” Series Bottle Vapor Trap and Indicator
Click to view available options
No. per Case:
5/Pk.
Each
Description
Not recommended for use under pressure with Luer products fitting as no Luer lock is employed.
Order Info
Available with Luer port or Luer port with fluorocarbon check valve and 10μm porosity PTFE inlet filter enabling air to flow into the bottle while preventing particulate contamination and preventing harmful vapors from escaping.
Specifications
Material | PTFE |
No. per Case | Each |
Neck Finish | 1/4-28 |
For Use With (Equipment) | For distilled water, foodstuff |