missing translation for 'onlineSavingsMsg'
Learn More

Omnifit™ SolventPlus™ Chromatography Columns and Replacement Parts

Designed for use with organic solvents but equally compatible with aqueous solvent systems

Brand:  Kinesis 006SCC-10-15-AF

Code : Z3

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 13184104

  • POA / Each

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Specifications

Omnifit SolventPlus chromatography columns
150mm
10mm
SolventPlus chromatography column
10mm
Glass column assembly, frit kit, fittings kit; One adjustable, one fixed endpiece
Videos