missing translation for 'onlineSavingsMsg'
Learn More

Omnifit™ SolventPlus™ Chromatography Columns and Replacement Parts

Designed for use with organic solvents but equally compatible with aqueous solvent systems

Brand:  Omnifit Labware 006SCC-06-10-FF

UNSPSC : 41115711

Code : Z3

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 13134094

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications Description & Specifications

Specifications

Omnifit SolventPlus chromatography columns
100mm
6.6mm
SolventPlus chromatography column
6.6mm
Glass column assembly, frit kit, fittings kit; Two fixed endpieces