Learn More
Kinesis™ Omnifit™ SolventPlus™ Chromatography Columns and Replacement Parts
Designed for use with organic solvents but equally compatible with aqueous solvent systems
Brand:  Kinesis™ 006SFE-15
Code : Z3
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 13184164
  • POA / Each
This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.
Description
Description
  • Borosilicate glass and PTFE construction, with Chemraz™ O-rings and PTFE frits put only inert materials in the fluid path
  • ETFE end caps and fitting nuts ensure a chemically inert connection into and out of the system
  • PTFE endpiece makes the primary seal to glass and has even distribution pattern to give even sample application across the bed surface
  • Ready-assembled column includes fixed or adjustable endpieces (-FF, -AF or -AA configuration) and 30μm PE frits
Specifications
Specifications
Omnifit SolventPlus chromatography columns
PTFE
15mm
Replacement fixed endpieces for SolventPlus column
15mm
Videos