Learn More
Kinesis™ Omnifit™ SolventPlus™ Chromatography Columns and Replacement Parts
Designed for use with organic solvents but equally compatible with aqueous solvent systems
Brand:  Kinesis™ 006SAE-06
Code : Z3
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 13134084
  • £243.00 / Each
Estimated Shipment
13-06-2018
Add to basket
Description
Description
  • Borosilicate glass and PTFE construction, with Chemraz™ O-rings and PTFE frits put only inert materials in the fluid path
  • ETFE end caps and fitting nuts ensure a chemically inert connection into and out of the system
  • PTFE endpiece makes the primary seal to glass and has even distribution pattern to give even sample application across the bed surface
  • Ready-assembled column includes fixed or adjustable endpieces (-FF, -AF or -AA configuration) and 30μm PE frits
Specifications
Specifications
Omnifit SolventPlus chromatography columns
Replacement adjustable endpieces for SolventPlus column
6.6mm
6.6mm
Videos