missing translation for 'onlineSavingsMsg'
Learn More

Diba Omnifit™ PTFE Cones

For use with Omnifit cap-style connectors (1/16 in. PTFE cone is supplied with all standard bore cap-style connectors and valves)

Brand:  Diba 001502

Code : Z3

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 12786138

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Match to the O.D. of tubing used
  • Compressed onto the tubing to form a seal
  • Resistant to most chemicals including sulphuric acid, sodium hydroxide, and all organic solvents
  • Enable very low dead volme fluid path
  • 50 psi (3bar) pressure rating
  • Other PTFE cones for different tubing sizes are available; contact your Customer Service Representative

Order Info

Other PTFE cones for different tubing sizes are available; contact your Customer Service Representative

Specifications

Specifications

0.0625 in. Tubing
50 psi (3 bar)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers