Learn More
Diba Omnifit™ Packing Sleeves for EZ and EZ SolventPlus™ Columns
Use Omnifit™ Packing Sleeves for EZ and EZ SolventPlus™ Columns to join two columns for filling and packing
Brand:  Diba 006EPS-35
Additional Details : Weight : 0.23000kg
 View more versions of this product
Product Code. 15865274
  • POA / Each
In Stock
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Specifications
35 mm
Omnifit™ EZ Packing Sleeve, Join 2 Columns or Connect to Column Packer, 35mm; 1/ea
Liquid Chromatography Column
Omnifit™ EZ and EZ SolventPlus™ Columns