Learn More
Kinesis™ Omnifit™ Chromatography Column Replacement Endpieces: Fixed
Brand:  Kinesis™ 006BFE-03-SS
Code : Z3
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 13184034
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
3mm
0.2μm
PTFE
BenchMark columns used in chromatography
Fixed Length
Videos