missing translation for 'onlineSavingsMsg'
Learn More

Kinesis™ Omnifit™ Chromatography Column Replacement Endpieces: Fixed

Brand:  Kinesis™ 006BFE-03-50

Code : Z3

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 13164034

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

0.11 in.
BenchMark columns used in chromatography
Fixed Length
3mm
50μm
PTFE
Videos