missing translation for 'onlineSavingsMsg'
Learn More

Diba Omnifit™ Chromatography Column Replacement Endpieces: Fixed

No need to worry about losing an endpiece

Brand:  Diba 006BFE-10

Code : Z3

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 13104044

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

The endpieces make the primary seal to glass microbore columns.
Specifications

Specifications

PE frit
1 cm
39.2°F to 140°F
25 μm
600 psi
1 to 14
Fixed Length
0.39 in.
600 psi
4°C to 60°C
0.25 in. to 28 flat bottom
BenchMark Columns Used in Chromatography
Fixed Endpiece
PTFE
Videos
SDS
Documents
Certificates