missing translation for 'onlineSavingsMsg'
Learn More

Omnifit Chromatography Column Replacement Endpieces: Adjustable

Brand:  Kinesis 006BAE-25

Code : Z3

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 13143954

  • POA / Each

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Specifications

PE frit
BenchMark columns used in chromatography
25μm
Adjustable Length
25mm
4 to 60°C
Adjustable endpiece
PTFE
Videos