missing translation for 'onlineSavingsMsg'
Learn More

Kinesis™ Omnifit™ BenchMark™ Replacement Frits for Chromatography Column Assemblies

Bed support suited to a variety of applications and solvents

Brand:  Kinesis™ 006FR-15-25-PE

Code : Z3

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 13101314

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

15mm
Polyethylene
25μm
Chromatography Column assemblies
Universal
Replacement chromatography frit
Videos