Learn More
Kinesis™ Omnifit™ BenchMark™ Replacement Endpieces for Chromatography Column Assemblies
Have a distribution pattern to give even application of sample across the bed surface
Brand:  Kinesis™ 006BFE-15
Code : Z3
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 13114044
  • £115.00 / Each
Estimated Shipment
18-06-2018
Add to basket
Description
Description
  • PTFE construction makes a primary seal to the glass column
Specifications
Specifications
Fixed Endpieces
15mm
Chromatography Column Assemby - Replacement Endpieces
Videos