Learn More
Kinesis™ Omnifit™ BenchMark™ Replacement Endpieces for Chromatography Column Assemblies
Have a distribution pattern to give even application of sample across the bed surface
Brand:  Kinesis™ 006BAE-50
Code : Z3
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 13163954
  • £538.00 / Each
Estimated Shipment
11-05-2018
Add to basket
Description
Description
  • PTFE construction makes a primary seal to the glass column
Specifications
Specifications
Adjustable endpieces
50mm
Chromatography Column Assemby - Replacement Endpieces
Videos