Learn More
Kinesis™ Omnifit™ BenchMark™ Microbore Chromatography Columns
Optimized for aqueous systems and compatible with solvents used in common liquid chromatography applications like protein purification
Brand:  Kinesis™ 006BCC-03-15-FF
Code : Z3
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 13103964
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
  • Economical, nonadjustable columns for low pressure or gravity feed
  • 3mm diameter and a variety of lengths
  • Compatible with a variety of bulk media
Not resistant to acetone, ketones, chlorinated hydrocarbons, aliphatic esters, phenol, >10% NaOH, >10% HCl, >5% acetic acid, or strong mineral acid.
Specifications
Specifications
150mm
3mm
Videos