missing translation for 'onlineSavingsMsg'
Learn More

Omnifit™ Benchmark™ Low-Pressure Chromatography Column Assemblies

Designed to suit the majority of standard laboratory chromatography applications, including protein purification

Brand:  Kinesis 006BCC-06-40-FF

Code : Z3

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 13133974

  • POA / Each

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Specifications

25μm
400mm
Borosilicate Glass
6.6mm
Videos