missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse OMgp Antibody, R&D Systems™

Rat Monoclonal Antibody

POA

Specifications

Antigen OMgp
Clone 240527
Concentration LYOPH
Dilution Western Blot 1 ug/mL
Applications Western Blot
View More Specs

Description

Description

OMgp Monoclonal specifically detects OMgp in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

OMgp
LYOPH
Western Blot
Unconjugated
Rat
Human, Mouse
Q63912
4974
Mouse myeloma cell line NS0-derived recombinant mouse OMgp Ile25-Ser420 Accession # Q63912
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human and mouse OMgp in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human OMgp is observed.
240527
Western Blot 1 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
oligodendrocyte myelin glycoprotein, OMG, OMGPoligodendrocyte-myelin glycoprotein
OMG
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Certificates

For Research Use Only