Learn More
Olvanil, Tocris Bioscience™
Potent vanilloid receptor agonist
Brand:  Tocris (Bio-Techne) 093450
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13488173
  • £427.72 / 50mg
Estimated Shipment
04-10-2018
Add to basket
Description
Description
Potent vanilloid receptor agonist (pEC50 values are 8.1 and 7.7 at rat and human VR1 receptors respectively). Also blocks anandamide uptake (IC50 = 9 μM) and may bind to CB1 cannabinoid receptors. Antinociceptive following systemic administration. Also available as part of the Vanilloid TRPV1 Receptor Tocriset™ .
Specifications
Chemical Identifiers
58493-49-5
417.634
N-Vanillyloleoylamide
(Z)-N-[(4-hydroxy-3-methoxyphenyl)methyl]octadec-9-enamide
C26H43NO3
OPZKBPQVWDSATI-KHPPLWFESA-N
5311093
CCCCCCCCC=CCCCCCCCC(=O)NCC1=CC(=C(C=C1)O)OC
Specifications
Olvanil
>98%
N-Vanillyloleoylamide
CCCCCCCCC=CCCCCCCCC(=O)NCC1=CC(=C(C=C1)O)OC
417.634
417.63
58493-49-5
C26H43NO3
OPZKBPQVWDSATI-KHPPLWFESA-N
(Z)-N-[(4-hydroxy-3-methoxyphenyl)methyl]octadec-9-enamide
5311093