missing translation for 'onlineSavingsMsg'
Learn More

Olomoucine, Tocris Bioscience™

Cyclin-dependent kinase inhibitor

Brand:  Tocris (Bio-Techne) 12845

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 5mg

 View more versions of this product

Product Code. 13459053

  • £117.45 / 5mg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

Cyclin-dependent kinase inhibitor that competes for the ATP binding site of the kinase. Selectively inhibits cdc2/cyclin B (IC50 = 7 μM), cdk2/cyclin A (IC50 = 7 μM), cdk2/cyclin E (IC50 = 7 μM), cdk/p35 kinase (IC50 = 3 μM)? and ERK1/MAP kinase (IC50 = 25 μM). Arrests human fibroblasts in the G1 phase.
Specifications

Chemical Identifiers

101622-51-9
298.35
olomoucine, 4erk, unii-6a839b2hys, insolution olomoucine, 2-2-hydroxyethylamino-6-benzylamino-9-methylpurine, 6-benzylamino-2-2-hydroxyethylamino-9-methylpurine, 6-benzylamino-2-2-hydroxyethylamino-9-methylpurine, 2-6-benzylamino-9-methyl-9h-purin-2-yl amino ethanol, ethanol,2-9-methyl-6-phenylmethyl amino-9h-purin-2-yl amino, 2-hydroxyethylamino-6-benzylamino-9-methylpurine
2-[[6-(benzylamino)-9-methylpurin-2-yl]amino]ethanol
C15H18N6O
GTVPOLSIJWJJNY-UHFFFAOYSA-N
4592
CN1C=NC2=C1N=C(N=C2NCC3=CC=CC=C3)NCCO

Specifications

Olomoucine
101622-51-9
olomoucine, 4erk, unii-6a839b2hys, insolution olomoucine, 2-2-hydroxyethylamino-6-benzylamino-9-methylpurine, 6-benzylamino-2-2-hydroxyethylamino-9-methylpurine, 6-benzylamino-2-2-hydroxyethylamino-9-methylpurine, 2-6-benzylamino-9-methyl-9h-purin-2-yl amino ethanol, ethanol,2-9-methyl-6-phenylmethyl amino-9h-purin-2-yl amino, 2-hydroxyethylamino-6-benzylamino-9-methylpurine
CN1C=NC2=C1N=C(N=C2NCC3=CC=CC=C3)NCCO
298.35
298.35
5mg
C15H18N6O
GTVPOLSIJWJJNY-UHFFFAOYSA-N
2-[[6-(benzylamino)-9-methylpurin-2-yl]amino]ethanol
4592
>98%
Videos