missing translation for 'onlineSavingsMsg'
Learn More

Olomoucine, 98%, ACROS Organics™

Brand:  Acros Organics 328410010

Code : 29

Additional Details : CAS Number : 101622-51-9 Weight : 0.05001kg

Quantity 1mg

 View more versions of this product

Product Code. 10046163

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Olomoucine
101622-51-9
100.0
98%
298.4
6-Benzylamino-2-(2-hydroxyethylamino)-9-methylpurine
98%
97.5
98%
C15H18N6O
MFCD00189360

Specifications

Olomoucine
101622-51-9
100.0
Solid
98%
298.4
6-Benzylamino-2-(2-hydroxyethylamino)-9-methylpurine
White to off-white
1mg
98%
97.5
Authentic
98%
C15H18N6O
MFCD00189360
Solubility in water: slightly soluble in water. Other solubilities: soluble in dmso and methanol
120.0° to 130.0°C
Videos
SDS