Learn More
Oligodendrocyte Marker O1 Monoclonal Antibody (O1), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-6506-82
Code : Z2
Additional Details : Weight : 0.01000kg
Description
Description: This O1 monoclonal antibody reacts with galactocerebroside and other lipids found on late oligodendrocyte precursors. O1 has been reported to react to all species tested (rat, mouse, human, chicken). Oligodendrocytes function as myelin-producing cells within the central nervous system ensheathing the axons of neurons with myelin in order to facilitate rapid axonal conduction. O1 and O4 monoclonal antibodies serve as stage-specific markers of oligodendrocyte development with O4 marking early oligodendrocyte progenitors and O1-labeling late oligodendrocyte progenitors. O4-positive cells differentiate to become O1-positive, which can in turn become mature oligodendrocytes. Applications Reported: This O1 antibody has been reported for use in flow cytometric analysis, immunohistochemical staining of frozen tissue sections, ELISA, and immunocytochemistry. Applications Tested: This O1 antibody has been tested by immunocytochemistry on formaldehyde-fixed, differentiated OLN93 cells and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Oligodendrocytes function as myelin-producing cells within the central nervous system ensheathing the axons of neurons with myelin in order to facilitate rapid axonal conduction. O1 and O4 monoclonal antibodies serve as stage-specific markers of oligodendrocyte development with O4 marking early oligodendrocyte progenitors and O1-labeling late oligodendrocyte progenitors. O4-positive cells differentiate to become O1-positive, which can in turn become mature oligodendrocytes.Specifications
Oligodendrocyte Marker O1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse | |
100 μg | |
Primary | |
4° C | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen) | |
O1 | |
Unconjugated | |
O1 Marker | |
Affinity chromatography | |
RUO | |
Chicken, Human, Mouse, Rat | |
Antibody | |
IgM |
For Research Use Only.