missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OLIG2 (Marker of Glial Brain Tumors) Mouse, Clone: OLIG2/2400, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 10215-MSM1-P1
Additional Details : Weight : 0.01000kg
Specifications
OLIG2 (Marker of Glial Brain Tumors) | |
Nuclear | |
OLIG2/2400 | |
Unconjugated | |
Mouse | |
30-40kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
10215 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment of human OLIG2 protein (apprx. aa 1-141) | |
THP-1 cells. Astrocytoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |