missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oleic Acid, Pure, Technical, Fisher Chemical™
Click to view available options
Quantity:
2.5 L
25 L
Packaging:
Amber glass bottle
Plastic container
Specifications
Melting Point | 13°C |
Color | Colorless |
Boiling Point | 360°C |
Packaging | Amber glass bottle |
Quantity | 2.5 L |
Vapor Density | 9.7 |
Viscosity | 39.1 mPaS at 20°C |
Formula Weight | 282.46g/mol |
Vapor Pressure | 1mmHg 176 |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'