Learn More
Oleamide, Tocris Bioscience™
Sleep-inducing brain lipid
Brand:  Tocris (Bio-Techne) 087810
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13488093
  • £73.40 / 10mg
Estimated Shipment
07-03-2018
Add to basket
Description
Description
Endogenous sleep-inducing lipid. Acts as an agonist at the CB1 cannabinoid receptor (EC50 = 1.64 μM). Also appears to potentiate the actions of 5-HT on 5-HT2A and 2C receptors, and act via an allosteric regulatory site on 5-HT7 receptors.
Specifications
Chemical Identifiers
Oleamide
C18H35NO
281.48
cis-9-Octadecenamide
Specifications
Oleamide
>98%
281.48
10mg
301-02-0
C18H35NO
cis-9-Octadecenamide
Videos