missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ OIML Class M1 Proof Line Weight Set without Certificate

Includes the same features and properties as the individual weights of this series, along with the corresponding maximum permissible errors. Sartorius™ OIML Class M1 Weight Set without Certificate is supplied with gloves, tweezers and brush in a high-quality plastic case

£138.00 - £940.00

Specifications

Product Type Proof Line Weight Set
Classification OIML Class M1
View More Specs

Products 3
Product Code Brand Mass Material Density Price Quantity & Availability  
Product Code Brand Mass Material Density Price Quantity & Availability  
11303364
View Documents Promotion Details
Sartorius™
YCS01-255-AC-00
1 mg to 500 mg Aluminum (1 to 5 mg), Nickel Silver (10 to 500 mg) 2,950 kg/m3 (1 to 5mg), 8,600 kg/m3 (10mg to 500mg)
£138.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-05-2024
to see stock.
Add to basket
 
11363374
View Documents Promotion Details
Sartorius™
YCS31-615-AC-00
1 g to 1 kg Stainless Steel 7,850 kg/m3
£426.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
 
11343384
View Documents Promotion Details
Sartorius™
YCS31-655-AC-00
1 g to 5 kg Stainless Steel 7,850 kg/m3
£940.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
 
Description

Description

Values 1 to 5mg: Aluminum leaf weights with density of 2,650 kg/m3
Values of 10 to 500mg: Nickel silver construction with density of 8,600 kg/m3.
Values of 1g and higher: Polished, special stainless steel knob weights with density of 7,950kg/m3
  • Highly corrosion resistant
  • Non-magnetizable
  • Tolerances comply with OIML R111-1
  • Includes sturdy plastic carry case
Specifications

Specifications

Proof Line Weight Set
OIML Class M1
Videos
SDS
Documents

Documents

Product Certifications