missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ OIML Class F2 Proof Line Knob Weight without Certificate
Polished non-magnetizable calibration weights. Sartorius™ OIML Class F2 Proof Line Knob Weight without Certificate is constructed of special steel and supplied in a plastic case.
£34.90 - £630.00
Specifications
Product Type | Proof Line Weight |
---|---|
Classification | OIML Class F2 |
Material | Special Stainless Steel |
Density | 7,950kg/m3 |
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
11363384
|
Sartorius™
YCW314-AC-00 |
1 g |
£34.90
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11303394
|
Sartorius™
YCW324-AC-00 |
2 g |
£36.70
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11343394
|
Sartorius™
YCW354-AC-00 |
5 g |
£37.60
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11323404
|
Sartorius™
YCW424-AC-00 |
20 g |
£43.10
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11363404
|
Sartorius™
YCW454-AC-00 |
50 g |
£45.90
Each |
Estimated Shipment: 24-04-2024
Log in to see stock. |
|||||
11303414
|
Sartorius™
YCW514-AC-00 |
100 g |
£51.40
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11343414
|
Sartorius™
YCW524-AC-00 |
200 g |
£68.90
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
11383414
|
Sartorius™
YCW554-AC-00 |
500 g |
£123.00
Each |
In Stock
Log in to see stock. |
|||||
11303434
|
Sartorius™
YCW654-AC-00 |
5 kg |
£321.00
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11387494
|
Sartorius™
YCW714-AC-00 |
10 kg |
£630.00
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11383394
|
Sartorius™
YCW414-AC-00 |
10 g |
£40.40
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11323424
|
Sartorius™
YCW614-AC-00 |
1 kg |
£131.00
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
Description
- Non-magnetizable
- Supplied in a high-quality plastic case (unless otherwise specified) suitable for use in cleanrooms
- Special Stainless steel construction with a density of 7,950 kg/m3
- Complies with OIML R111-1 International Recommentation
Specifications
Proof Line Weight | |
OIML Class F2 | |
Special Stainless Steel | |
7,950kg/m3 |